Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thalidomide-1-Me,4-OH | 2244568-06-5 | ≥98.0% | 288.26 | C14H12N2O5 | 50 MG
SDP

Supplier:  Medchemexpress LLC HYW44137650MG

Encompass_Preferred

Thalidomide-1-Me,4-OH is a thalidomide analogue bearing a 4-hydroxy substituent and a 1-methyl modification. It serves as a cereblon/E3 ligase ligand and is used as a recruiter in PROTAC and targeted protein degradation research. Supplied as a research-grade chemical for biochemical and cellular studies.

  • Ligand for E3 ligase (cereblon) used in PROTAC applications.
  • Thalidomide derivative with 4-hydroxy and 1-methyl modifications.
  • Molecular formula C14H12N2O5; molecular weight 288.26.
  • Purity ≥98.0% suitable for research use.
  • Storage guidance: powder -20°C (3 years), 4°C (2 years).

Catalog No. 50-002-81444


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.