Learn More
Medchemexpress LLC Thalidomide 4-chloro | 244057-36-1 | MFCD30188071 | 96.0% | 292.68 g·mol⁻¹ | C13H9ClN2O4 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYW460141500MG
Encompass_Preferred
Thalidomide 4-chloro is a chlorinated derivative of thalidomide that acts as an E3 ligase activator for biochemical and cell-based research. It is commonly used as a research reagent and as a synthetic building block for further derivatization at the 4-chloro position, and is offered in lab-scale pack sizes with supporting product documentation.
  • Acts as an E3 ligase activator for protein degradation studies.
  • Serves as a synthetic intermediate for derivatization at the 4-chloro position.
  • Applicable in biochemical and cell-based assays.
  • Supplied in multiple lab-scale pack sizes suitable for research.
  • Molecular identity confirmed: CAS 244057-36-1, formula C13H9ClN2O4, MW 292.68 g·mol⁻¹.
Catalog No. 50-002-81093