Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thalidomide 4-fluoride | 835616-60-9 | 99.9% | 276.22 | 5 G
SDP

Supplier:  Medchemexpress LLC HY415475G

Encompass_Preferred

Thalidomide 4-fluoride is a Thalidomide-based Cereblon ligand that recruits CRBN protein and can be linked to the ligand for IRAK4 protein to create PROTAC IRAK4 degrader-1. This compound targets cereblon (CRBN), a ubiquitous protein involved in the cullin-4 E3 ubiquitin ligase complex CRL4, facilitating the ubiquitination and subsequent proteasomal degradation of target proteins.

  • Recruits CRBN protein.
  • Can be linked to IRAK4 protein ligand.
  • Targets cereblon (CRBN).
  • Facilitates ubiquitination and proteasomal degradation of target proteins.
  • Molecular weight: 276.22.
  • Physical state: solid.
  • Color: off-white to gray.
  • Soluble in DMSO.
  • Store powder at -20°C for 3 years or 4°C for 2 years.
  • Store in solvent at -80°C for 2 years or -20°C for 1 year.
  • Avoid repeated freeze-thaw cycles after preparation into solution.

Catalog No. 50-004-04911


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.