Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thalidomide-4-O-C10-COOH | 2379870-45-6 | MFCD03424822 | 99.1% | 458.50 g·mol⁻¹ | C24H30N2O7 | 5 MG
SDP

Supplier:  Medchemexpress LLC HY1328585MG

Encompass_Preferred

Thalidomide-4-O-C10-COOH is an E3 ligase ligand-linker conjugate intended as a building block for PROTAC synthesis. It features a thalidomide-derived cereblon-recruiting motif linked via a C10 spacer to a terminal carboxylic acid, allowing standard coupling chemistry for attachment to target-binding ligands. Supplied as a high-purity solid for laboratory research use.

  • High purity suitable for synthesis work.
  • Carboxylic acid terminus for standard coupling reactions.
  • Contains a C10 spacer to provide linker flexibility.
  • Molecular formula: C24H30N2O7; molecular weight: 458.50 g·mol⁻¹.
  • Supplied in small mg-scale quantities appropriate for research.

Catalog No. 50-002-62742


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.