Learn More
Medchemexpress LLC Thalidomide-4-o-c12-nh2 hydrochloride | 95.6% | 494.02 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY149574
Encompass_Preferred
Thalidomide-4-O-C12-NH2 hydrochloride is a thalidomide-based cereblon ligand utilized for the recruitment of CRBN protein. It can be linked to a protein ligand via a linker to form PROTACs.
  • Thalidomide-based cereblon ligand
  • Utilized for recruitment of CRBN protein
  • Can be linked to a protein ligand via a linker to form PROTACs
  • Target: Cereblon
Catalog No. 50-003-61065