Learn More
Medchemexpress LLC Thalidomide 5-fluoride | 835616-61-0 | 276.22 g/mol | C13H9FN2O4 | 5 G

Supplier: Medchemexpress LLC HYW0873835G
Thalidomide 5-fluoride is a 5-fluoro derivative of thalidomide that functions as a cereblon (CRBN) ligand for use in the design and synthesis of PROTACs and other E3 ligase-recruiting molecules.
- Used as a cereblon ligand in PROTAC and degrader synthesis.
- CAS number 835616-61-0.
- Molecular formula C13H9FN2O4.
- Molecular weight 276.22 g/mol.
- Packaged as a 5 g quantity.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.