Learn More
Medchemexpress LLC Thalidomide-5-NH2-C8-NH2 TFA | 2097509-49-2 | 514.49 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1488725MG
Encompass_Preferred
Thalidomide-5-NH2-C8-NH2 TFA is a chemical compound primarily used for compound synthesis and various biochemical assay applications. It is characterized by its specific molecular weight and solubility properties.
  • Used in compound synthesis
  • Suitable for biochemical assay reagents
  • Soluble in DMSO (up to 100 mg/mL with ultrasonic assistance)
  • Requires sealed storage at -20°C, away from moisture and light
  • When in solvent, store at -80°C for up to 6 months, or -20°C for up to 1 month, under sealed conditions away from moisture and light.
Catalog No. 50-003-61968