Learn More
Medchemexpress LLC Thalidomide-5-O-C10-NH2 hydrochloride | 99.4% | 465.97 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY149567
Encompass_Preferred
Thalidomide-5-O-C10-NH2 hydrochloride is a Thalidomide-based cereblon ligand. It is used to recruit CRBN protein and can be linked to a protein ligand via a linker to form PROTACs. This product is for research use in laboratory settings and for the manufacture of substances.
  • Cereblon ligand
  • Recruits CRBN protein
  • Forms PROTACs with protein ligands
  • For research use only
  • Appearance: Solid
  • Color: White to off-white
Catalog No. 50-003-63964