Learn More
Medchemexpress LLC Thalidomide-5-O-C10-NH2 hydrochloride | 99.4% | 465.97 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1495675MG
Encompass_Preferred
Thalidomide-5-O-C10-NH2 hydrochloride is a cereblon ligand based on Thalidomide and is utilized in the recruitment of CRBN protein. It can be linked to a target protein ligand via a linker to form PROTACs.
  • Functions as a ligand for E3 ligase, specifically cereblon.
  • Utilized in the formation of PROTACs by connecting to a protein ligand via a linker.
  • Intended for research use only and not for sale to patients.
Catalog No. 50-003-60319