Learn More
Medchemexpress LLC Thalidomide-5-O-C9-NH2 hydrochloride | 99.6% | 451.94 | 25 MG

Supplier: Medchemexpress LLC 25MGHY149566
Thalidomide-5-O-C9-NH2 hydrochloride is a thalidomide-based cereblon ligand used in the recruitment of CRBN protein. It can be connected to a ligand for protein by a linker to form PROTACs.
- Cereblon ligand
- Forms PROTACs
- Recruits CRBN protein
- Target: Cereblon
- Molecular formula: C22H30ClN3O5
- Appearance: Solid
- Color: White to off-white
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.