Learn More
Medchemexpress LLC Thalidomide-5-PEG2-Cl | 2230956-57-5 | 96.57% | 380.78 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY139543100MG
Encompass_Preferred
Thalidomide-5-PEG2-Cl is a Thalidomide-based cereblon ligand involved in the recruitment of CRBN protein. It can be connected to a protein's ligand via a linker to form PROTACs. PROTACs utilize two different ligands connected by a linker: one binds to an E3 ubiquitin ligase, and the other targets the desired protein, leveraging the intracellular ubiquitin-proteasome system for selective degradation of target proteins.
  • Utilized in the recruitment of CRBN protein
  • Can be linked to a protein's ligand to form PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Enables selective degradation of target proteins
  • Functions as an E3 ligase ligand-linker conjugate
Catalog No. 50-003-50222