Learn More
Medchemexpress LLC Thalidomide-5-peg2-cl | 2230956-57-5 | 96.6% | C17H17ClN2O6 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13954310MG
Encompass_Preferred
Thalidomide-5-PEG2-Cl is a thalidomide-derived cereblon (CRBN) ligand featuring a short PEG2 spacer and a terminal chloride. It is supplied as a research-grade building block for conjugation in PROTAC design and other CRBN-targeting chemical biology applications, with documented molecular formula and purity.
  • Provides a CRBN-recruiting moiety suitable for PROTAC assembly.
  • Contains a PEG2 linker for flexible conjugation to ligands.
  • Terminal chloride enables straightforward chemical derivatization.
  • Supplied with referenced purity and characterization data.
  • Available in small research-friendly pack sizes for early-stage work.
Catalog No. 50-002-59624