Learn More
Medchemexpress LLC Thalidomide-5-PEG4-NH2 hydrochloride | 2743434-24-2 | 95.2% | 485.92 | 50 MG
Supplier: Medchemexpress LLC HY138785A50MG
Thalidomide-5-PEG4-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate. It incorporates a Thalidomide-based cereblon ligand and a linker, and is used in PROTAC technology. This product is intended for research use only and is not sold to patients.
- Synthesized E3 ligase ligand-linker conjugate
- Incorporates Thalidomide-based cereblon ligand
- Utilizes a linker for PROTAC technology
- For research use only