Learn More
Medchemexpress LLC Thalidomide-C2-amido-C2-COOH | 2353496-84-9 | 98.45% | 50 MG
SDP
Supplier:  Medchemexpress LLC HY13071350MG
Encompass_Preferred
Thalidomide-C2-amido-C2-COOH incorporates a CRBN ligand for the E3 ubiquitin ligase and a linker. It can be used to design PROTAC CDK2/9 Degrader-1 (HY-130709).
  • Incorporates a CRBN ligand for the E3 ubiquitin ligase and a linker.
  • Used to design PROTAC CDK2/9 degrader-1.
  • For research use only.
Catalog No. 50-003-39838