Learn More
Medchemexpress LLC Thalidomide-CH2NH2 hydrochloride | 444287-40-5 | 98.1% | 323.73 | 100 MG

Supplier: Medchemexpress LLC HY44249100MG
Thalidomide-CH2NH2 hydrochloride is a Thalidomide analogue featuring a primary amine, a versatile functional group that can participate in various reactions. It is intended for research use only.
- Features a primary amine group
- Provided as a solid, white to off-white in color
- Ships at room temperature in continental US
- Powder storage at -20°C for 3 years, 4°C for 2 years
- Storage in solvent at -80°C for 6 months, -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.