Learn More
Medchemexpress LLC Thalidomide-CH2NH2 hydrochloride | 444287-40-5 | 98.1% | 323.73 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY4424950MG
Encompass_Preferred
Thalidomide-CH2NH2 hydrochloride is a Thalidomide analogue featuring a primary amine, which is a versatile group that may participate in many reactions. It is for research use only.
  • Features a primary amine
  • Versatile group that may participate in many reactions
  • For research use only
Catalog No. 50-004-05122