Learn More
Medchemexpress LLC Thalidomide-NH-amido-PEG4-C2-NH2 hydrochloride | 2983037-00-7 | 98.0% | C25H36ClN5O9 | 10MG
SDP
Supplier:  Medchemexpress LLC HY138857A10MG
Encompass_Preferred
Thalidomide-NH-amido-PEG4-C2-NH2 hydrochloride is a thalidomide-based cereblon (CRBN) ligand-linker conjugate intended as a building block for PROTAC synthesis. The molecule combines a thalidomide-derived CRBN recruiting moiety with an amino-terminated PEG4-C2 spacer to enable straightforward coupling to target ligands. It is supplied as the hydrochloride salt for improved handling and is for research use only.
  • High HPLC purity (98.0%).
  • Amino-terminated PEG4-C2 linker for facile conjugation.
  • Thalidomide-based cereblon ligand for CRBN recruitment.
  • Supplied as a solid hydrochloride salt, light yellow to yellow.
  • Available in small-scale quantities suitable for medicinal chemistry.
  • Store sealed, away from moisture; in solvent: -80°C for 6 months, -20°C for 1 month.
Catalog No. 50-002-60641