Learn More
Medchemexpress LLC Thalidomide-NH-C5-NH2 hydrochloride | 2375194-03-7 | 99.86% | 394.85 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY136237
Encompass_Preferred
Thalidomide-NH-C5-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide based cereblon ligand and a linker used in PROTAC technology. This product is for research use only.
  • Purity of 99.86%
  • Inhibits Cereblon
  • Utilizes PROTAC technology for targeted protein degradation
Catalog No. 50-003-51873