Learn More
Medchemexpress LLC Thalidomide-o-amido-C3-COOH | 2308035-51-8 | 98.4% | 417.37 | C19H19N3O8 | 2 G

Supplier: Medchemexpress LLC HY1036122G
Thalidomide-O-amido-C3-COOH is an E3 ligase ligand-linker conjugate that combines a thalidomide-derived cereblon ligand with a short C3 linker terminating in a carboxylic acid. It is intended for use as a CRBN-recruiting building block in PROTAC synthesis and related targeted protein degradation research.
- used as a cereblon (CRBN) recruiting building block in PROTAC synthesis
- terminates in a carboxylic acid for conjugation to target ligands
- reported purity 98.43% by LCMS
- molecular weight 417.37; formula C19H19N3O8
- storage: 4°C under nitrogen; in solution store at -80°C (6 months) or -20°C (1 month)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.