Learn More
Medchemexpress LLC HY-131646 25mg Medchemexpress, Thalidomide-O-amido-C3-PEG3-C1-NH2 CAS:1799711-29-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13164625MG
Encompass_Preferred
Medchemexpress, HY-131646 25mg Thalidomide-O-amido-C3-PEG3-C1-NH2 CAS:1799711-29-7 Thalidomide-O-amido-C3-PEG3-C1-NH2 is a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide based cereblon ligand and 3-unit PEG linker used in PROTAC technology. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1291