Learn More
Medchemexpress LLC Thalidomide-o-amido-c3-peg3-c1-nh2 trifluoroacetate | 1799711-29-7 | MFCD32878273 | 99.1% | C27H35F3N4O11 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13164610MG
Encompass_Preferred
Thalidomide-O-amido-C3-PEG3-C1-NH2 is a synthesized E3 ligase ligand-linker conjugate that couples a thalidomide-derived cereblon ligand to a three-unit polyethylene glycol linker, designed for PROTAC synthesis and related chemical biology research. The compound is supplied as an off-white to light yellow oil, with molecular formula C27H35F3N4O11, molecular weight 648.58 g·mol⁻¹, and high purity suitable for research.
  • Contains a thalidomide-derived cereblon ligand for E3 ligase recruitment.
  • Includes a three-unit polyethylene glycol linker to enhance flexibility and solubility.
  • Designed for PROTAC design and targeted protein degradation studies.
  • High purity (99.1%) for consistent experimental results.
  • Characterized molecular properties facilitate reproducible use in research.
  • Available in small packaging sizes suitable for laboratory studies.
Catalog No. 50-002-61112