Learn More
Medchemexpress LLC Thalidomide-O-amido-C4-N3 | 2098488-36-7 | MFCD31560486 | 98.4% | 428.40 g/mol | C19H20N6O6 | 10 MG

Supplier: Medchemexpress LLC HY10361510MG
Thalidomide-O-amido-C4-N3 is a thalidomide-derived cereblon ligand-linker conjugate used as a building block in proteolysis-targeting chimera (PROTAC) and E3 ligase ligand research. It is provided with analytical characterization and storage guidance for research applications.
- Used as a cereblon ligand-linker conjugate for PROTAC and E3 ligase research.
- Molecular formula C19H20N6O6; molecular weight 428.40 g/mol.
- CAS number 2098488-36-7 for unambiguous identification.
- Purity 98.4% by LCMS (as supplied).
- Supplied in small research quantities (for example, 10 mg) and as stock solution preparations.
- Soluble in DMSO and compatible solvent systems for formulation.
- Store at 4°C under nitrogen; in solution, store at -80°C (6 months) or -20°C (1 month).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.