Learn More
Medchemexpress LLC Thalidomide-O-amido-C6-NH2 (hydrochloride) | 2376990-31-5 | 99.3% | 466.92 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY112618B
Encompass_Preferred
Thalidomide-O-amido-C6-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate that integrates the Thalidomide-based cereblon ligand with a linker, making it suitable for synthesizing PROTACs. PROTACs utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins by connecting an E3 ubiquitin ligase ligand with a ligand for the target protein via a linker.
  • Synthesized E3 ligase ligand-linker conjugate.
  • Incorporates Thalidomide based cereblon ligand and a linker.
  • Can be used in the synthesis of PROTACs.
  • PROTACs exploit the ubiquitin-proteasome system for selective protein degradation.
  • For research use only.
Catalog No. 50-004-39859