Learn More
Medchemexpress LLC Thalidomide-O-amido-CH2-PEG3-CH2-NH-Boc | 1799711-31-1 | 98.4% | C30H42N4O11 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14517750MG
Encompass_Preferred
Thalidomide-O-amido-CH2-PEG3-CH2-NH-Boc is a synthesized E3 ligase ligand-linker conjugate. It incorporates a thalidomide based cereblon ligand and a linker. This product can be used for the synthesis of PROTAC BET degrader.
  • E3 ligase ligand-linker conjugate
  • Thalidomide based cereblon ligand
  • PROTAC BET degrader synthesis
  • Inhibitor
Catalog No. 50-003-55424