Learn More
Medchemexpress LLC Thalidomide-O-amido-PEG2-C2-NH2 (TFA) | 1957235-75-4 | 576.48 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY112617A
Encompass_Preferred
Thalidomide-O-amido-PEG2-C2-NH2 TFA is a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide-based cereblon ligand and 2-unit PEG linker used in PROTAC technology. PROTACs contain two different ligands connected by a linker, one for an E3 ubiquitin ligase and the other for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • E3 ligase ligand-linker conjugate
  • Incorporates thalidomide-based cereblon ligand
  • 2-unit PEG linker
  • Used in PROTAC technology
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-004-39980