Learn More
Medchemexpress LLC Thalidomide-O-C6-COOH | 2169266-69-5 | MFCD32063464 | 99.3% | 402.40 g·mol⁻¹ | C20H22N2O7 | 5 MG

Supplier: Medchemexpress LLC HY1309515MG
Thalidomide-O-C6-COOH is a thalidomide-based cereblon ligand-linker conjugate developed for use in targeted protein degradation (PROTAC) research. The molecule incorporates a thalidomide-derived cereblon-binding moiety tethered to a six-carbon linker ending in a carboxylic acid, enabling conjugation to target-binding ligands. It is supplied with high purity and characterized solubility and storage parameters for research use.
- Thalidomide-based cereblon ligand-linker conjugate suitable for PROTAC synthesis.
- Six-carbon linker terminating in a carboxylic acid for facile conjugation.
- Molecular weight 402.40 g·mol⁻¹ and formula C20H22N2O7.
- Purity 99.29% as supplied by the manufacturer.
- Soluble ≥ 100 mg/mL (248.51 mM) in DMSO.
- Supplied as an off-white to light yellow solid with recommended storage conditions.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.