Learn More
Medchemexpress LLC Thalidomide-O-C8-Boc | 2225148-52-5 | 95.1% | 50 MG
SDP
Supplier:  Medchemexpress LLC HY13115950MG
Encompass_Preferred
Thalidomide-O-C8-Boc is a thalidomide-based cereblon ligand that is used to recruit the CRBN protein. It can be linked to a protein ligand by a linker to create PROTACs. This product is intended for research use only.
  • Functions as a thalidomide-based cereblon ligand
  • Utilized for recruitment of CRBN protein
  • Connects to protein ligands via a linker to form PROTACs
  • Targets cereblon
Catalog No. 50-003-41169