Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thalidomide-O-COOH | 1061605-21-7 | MFCD31560477 | 99.9% | 332.27 g·mol⁻¹ | C15H12N2O7 | 10 MG
SDP

Supplier:  Medchemexpress LLC HY103597250MG

Encompass_Preferred

Thalidomide-O-COOH is a carboxylic acid-functionalized thalidomide derivative that serves as a cereblon (CRBN) ligand for targeted protein recruitment. It is used as an E3 ligase recruiter in PROTAC design and related chemical biology research.

  • Carboxylic acid-functionalized thalidomide cereblon ligand.
  • Used as an E3 ligase recruiter in PROTAC design and degrader development.
  • Molecular formula C15H12N2O7, molecular weight 332.27 g·mol⁻¹.
  • CAS number 1061605-21-7.
  • For research use only; not for human or diagnostic applications.

Catalog No. 50-002-45003


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.