Learn More
Medchemexpress LLC HY-126458 25mg Medchemexpress, Thalidomide-O-PEG2-propargyl CAS:2098487-52-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY12645825MG
Encompass_Preferred
Medchemexpress, HY-126458 25mg Thalidomide-O-PEG2-propargyl CAS:2098487-52-4 Thalidomide-O-PEG2-propargyl (E3 ligase Ligand-Linker Conjugates 32) is a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide based cereblon ligand and 2-unit PEG linker used in PROTAC technology. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1106