Learn More
Medchemexpress LLC Thalidomide-O-PEG4-amine (hydrochloride) | 2820929-03-9 | C23H32ClN3O9 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY141010A50MG
Encompass_Preferred
Thalidomide-O-PEG4-amine hydrochloride is a synthesized E3 ligase ligand-linker conjugate. It incorporates the Thalidomide based cereblon ligand and a linker used in PROTAC technology. This conjugate exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Incorporates thalidomide based cereblon ligand
  • Uses a linker in PROTAC technology
  • Targets E3 ligase ligand-linker conjugates, apoptosis, and autophagy
Catalog No. 50-003-55128