Learn More
Medchemexpress LLC Thalidomide-PEG4-COOH | 2828438-28-2 | 95.36% | 478.45 | 100 MG

Supplier: Medchemexpress LLC 100MGHY138774
Thalidomide-PEG4-COOH is a synthesized E3 ligase ligand-linker conjugate incorporating a Thalidomide-based cereblon ligand and a linker used in PROTAC technology. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is for research use only.
- Synthesized E3 ligase ligand-linker conjugate
- Incorporates a Thalidomide-based cereblon ligand
- Used in PROTAC technology for targeted protein degradation
- Exploits the intracellular ubiquitin-proteasome system
- Targets cereblon
- Appearance is oil
- Store at -20°C, protected from light
- Soluble in DMSO at 250 mg/mL
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.