Learn More
Medchemexpress LLC Thalidomide-PEG4-COOH | 2828438-28-2 | 95.4% | 478.45 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY13877450MG
Encompass_Preferred
Thalidomide-PEG4-COOH is a synthesized E3 ligase ligand-linker conjugate that incorporates a Thalidomide-based cereblon ligand and a linker used in PROTAC technology. This product is intended for research use only.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates thalidomide-based cereblon ligand
  • Utilizes a linker in PROTAC technology
  • For research use only
  • Appears as an oil
  • Color is colorless to light yellow
Catalog No. 50-003-50491