Learn More
Medchemexpress LLC Thalidomide-Piperazine 5-fluoride | 2222114-22-7 | 96.5% | 360.34 | 1 G
SDP
Supplier:  Medchemexpress LLC 1GHY134983
Encompass_Preferred
Thalidomide-Piperazine 5-fluoride is a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide based cereblon ligand and a linker used in PROTAC technology.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates Thalidomide based cereblon ligand
  • Utilizes linker in PROTAC technology
  • For research use only
  • Recommended storage at 4°C, protected from light
  • Targets E3 Ligase Ligand-Linker Conjugates, Apoptosis, and Autophagy pathways
  • Appears as a light yellow to green yellow solid
Catalog No. 50-003-65449