Learn More
Medchemexpress LLC Thalidomide-piperazine-peg1-cooh | 2731007-11-5 | 99.2% | 458.46 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY13878250MG
Encompass_Preferred
Thalidomide-Piperazine-PEG1-COOH is a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide based cereblon ligand and a linker used in PROTAC technology.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates thalidomide-based cereblon ligand
  • Used in PROTAC technology
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-49581