Learn More
Medchemexpress LLC Thalidomide-propargyl | 2098487-39-7 | 99.97% | 312.28 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY138776
Encompass_Preferred
Thalidomide-propargyl is a Thalidomide-based Cereblon ligand used for the recruitment of CRBN protein. It can be linked to a protein ligand via a linker to form IMiD-containing PROTACs. This compound is also a click chemistry reagent, featuring an Alkyne group that can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Cereblon ligand
  • Recruits CRBN protein
  • Forms IMiD-containing PROTACs
  • Click chemistry reagent
  • Features an Alkyne group
  • Participates in copper-catalyzed azide-alkyne cycloaddition (CuAAc)
Catalog No. 50-003-48496