Learn More
Medchemexpress LLC Thalidomide-pyrrolidine hydrochloride | 2616553-35-4 | 98.0% | 335.74 | C15H14ClN3O4 | 5 MG

Supplier: Medchemexpress LLC HYW248665A5MG
Thalidomide-pyrrolidine hydrochloride is a cereblon-binding E3 ligase ligand supplied as the hydrochloride salt for use as a building block in PROTAC synthesis and targeted protein degradation research. It is intended for research use and is accompanied by quality documentation.
- Cereblon-binding E3 ligase ligand for PROTAC synthesis.
- Hydrochloride salt form for handling and stability.
- High purity: 98.0%.
- Documented quality: data sheet, certificate of analysis, and safety data sheet available.
- Storage recommendations: store dry at room temperature; in solvent, -80°C (2 years) or -20°C (1 year).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.