Learn More
Medchemexpress LLC Thalidomide (standard) | 50-35-1 | 99.96% | 258.23 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY14658R
Encompass_Preferred
Thalidomide (Standard) is the analytical standard of Thalidomide, used for research and analytical applications. It inhibits cereblon (CRBN) with a Kd of approximately 250 nM, and has immunomodulatory, anti-inflammatory, and anti-angiogenic cancer properties. It can also function as a molecular glue to potentiate substrates.
  • Analytical standard grade
  • For research and analytical applications
  • Inhibits cereblon (CRBN) with Kd of approximately 250 nM
  • Immunomodulatory, anti-inflammatory, and anti-angiogenic cancer properties
  • Functions as a molecular glue to potentiate substrates
  • Used in qualitative, quantitative, and methodological research experiments (HPLC, GC, MS)
Catalog No. 50-003-63977