Learn More
Strem, An Ascensus Company CAS# 60969-19-9. 25g. Thallium(I) hexafluorophosphate, min. 97%. MFCD00049807
SDP
Supplier:  Strem, An Ascensus Company 81300025G
Encompass
CAS# 60969-19-9. 25g. Thallium(I) hexafluorophosphate, min. 97%. MFCD00049807. Molecular Weight: 349.34. Molecular Formula: TlPF6. Color/form: white xtl. Strem# 81-3000. http://www.strem.com/catalog/v/81-3000/
Catalog No. 50-901-13490