Learn More
Strem, An Ascensus Company CAS# 73491-36-8. 1g. Thallium(I) trifluoromethanesulfonate, 99% (Thallium triflate). MFCD00061608
SDP
Supplier:  Strem, An Ascensus Company 8185001G
Encompass
CAS# 73491-36-8. 1g. Thallium(I) trifluoromethanesulfonate, 99% (Thallium triflate). MFCD00061608. Molecular Weight: 353.45. Molecular Formula: TlSO3CF3. Color/form: white pwdr. Strem# 81-8500. http://www.strem.com/catalog/v/81-8500/
Catalog No. 50-901-13502