Learn More
Radiometer America Radiometer S7770 tHB Calibrating Solution for ABL90 FLEX PLUS
Supplier: Radiometer America 944021
S7770 tHB Calibrating Solution
S7770 tHB Calibrating Solution for the ABL90 FLEX PLUS blood gas analyzer
ABL90 FLEX PLUS Blood Gas Analyzer
The ABL90 FLEX PLUS analyzer gives you reliable results. With the ABL90 FLEX analyzer you get lab quality test results. In just 35 seconds, our cassette-based ABL90 FLEX analyzer can deliver 17 parameters from a sample as small as 65 μL. ABL90 measures blood gas, electrolytes, metabolites and co-oximetry. The ABL90 analyzer can measure a sample from a syringe, capillary tube or test tube.