Learn More
Medchemexpress LLC Thrombospondin-1 protein, mouse, recombinant, His-tagged | >95.0% | 10UG

Supplier: Medchemexpress LLC HYP70132510UG
Recombinant mouse thrombospondin-1 (THBS1), expressed in E. coli and carrying an N-terminal 6xHis tag, supplied lyophilized for research use. Corresponds to residues N19-K350 (accession Q80YQ1). Suitable for biochemical assays, antibody validation, and studies of cell-matrix interactions and angiogenesis.
- Expression system: E. coli
- N-terminal 6xHis tag
- Sequence range N19-K350 (accession Q80YQ1)
- Purity greater than 95% as determined by reducing SDS-PAGE
- Molecular weight approximately 36-40.6 kDa
- Lyophilized formulation in 10 mM Tris-HCl, 1 mM EDTA, 6% trehalose, pH 8.0
- Store at -20°C; after reconstitution stable at 4°C for 1 week
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.