Learn More
Boca Scientific Inc The Blocking Solution, 500ml
Supplier:  Boca Scientific Inc 110 500
Encompass
The Blocking Solution is a high-performance blocking reagent designed for saturating free binding sites on plastic surfaces or other protein binding surfaces and for minimizing nonspecific binding to surfaces in immunoassays. It is ideal for assays that require high reproducibility and low coefficients of variation, such as assays for pre-clinical and clinical studies, production of high quality diagnostics or measurement of precious specimens. The Blocking Solution is based on highly purified casein that has been chemically modified for maximum adsorption efficiency and a dense blocking layer. By incubating a surface with The Blocking Solution, such as a microtiter plate or western blotting membrane, free spaces on the surface are saturated by the components of this blocker. This minimizes unwanted binding of the analyte or the detection antibody to the surface, avoiding background and improving sensitivity of the assay.
Catalog No. NC0249376