Learn More
Bachem Miraculin (1-20)
SDP
Supplier:  Bachem 4016657.0005
Encompass_Preferred
Miraculin (1-20), H-Asp-Ser-Ala-Pro-Asn-Pro-Val-Leu-Asp-Ile-Asp-Gly-Glu-Lys-Leu-Arg-Thr-Gly-Thr-Asn-OH, 198694-37-0, C88H146N26O34, Mr 2112.28, 5mg. The eicosapeptide DSAPNPVLDIDGEKLRTGTN corresponds to the N-terminal sequence of the taste-modifying 191 amino acid-containing protein miraculin produced by the berries of Richadella dulcifica.
Catalog No. 50-194-6915