Learn More
Perkin Elmer US LLC Plastic Push-to-Connect Elbow Union for 1/4 O.D. Tubing
Supplier:  Perkin Elmer US LLC 09200001
Encompass_Preferred
The fitting has an all resin body material that is copper free. It is compatible with 1/4 O.D. tubing made from FEP, PFA, Nylon, Soft Nylon, or Polyurethane. It has an operating pressure range from -6 kPa to 1 Mpa (145 psi max.) and was designed to be used with either air or water applications.
Catalog No. 50-201-6556