Learn More
Bachem Suc-Leu-Leu-Val-Tyr-AMC
SDP
Supplier:  Bachem 4011369.0025
Encompass_Preferred
Suc-Leu-Leu-Val-Tyr-AMC, 94367-21-2, C40H53N5O10, Mr 763.89, 25mg. Chymotrypsin Substrate III, Fluorogenic. The fluorogenic substrate Suc-LLVY-AMC is used for measuring the chymotrypsin-like activity of the 20S proteasome, in combination with e.g. Bz-VGR-AMC (for assaying its trypsin-like activity) and Z-LLE-AMC (for determining its PGPH activity). Suc-LLVY-AMC is also cleaved by a chymotrypsin-like enzyme from sperm extract of the ascidian Halocynthia roretzi, a strong inhibitor of fertilization. The substrate has also been used to analyze the chymotrypsin-like activity of trypsins as well as chymase from rat mast cells. Furthermore, Suc-LLVY-AMC is a substrate for carboxypeptidase Y, proteinase yscE (kexin), ingensin, as well as for porcine calpain isozymes I and II and for papain.
Catalog No. 50-194-7009