Learn More
Bachem (Asp37)-Amyloid Beta Protein (1-42) Trifluoroacetate
SDP
Supplier:  Bachem 4099631.1000
Encompass_Preferred
(Asp37)-Amyloid Beta Protein (1-42) Trifluoroacetate, H-Asp-Ala-Glu-Phe-Arg-His-Asp-Ser-Gly-Tyr-Glu-Val-His-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Glu-Asp-Val-Gly-Ser-Asn-Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met-Val-Asp-Gly-Val-Val-Ile-Ala-OH, 1875128-79-2, C205H313N55O62S, Mr 4572.14, 1mg. [Asp37] - beta - Amyloid (1 - 42), [Asp37]-Abeta (1-42), Human, Amyloid-beta Peptide(1-42), G37D, Human. The G37D mutant does not show the aggregation behavior of WT Abeta42 nor its neurotoxicity.
Catalog No. 50-194-6592