Learn More
Bachem Fmoc-Dab(ivDde)-OH
SDP
Supplier:  Bachem 4066591.0005
Encompass_Preferred
Fmoc-Dab(ivDde)-OH, 607366-21-2, C32H38N2O6, Mr 546.66, 5g. The ivDde group can be cleaved selectively with 2% hydrazine hydrate in DMF. It is more stable towards the repetitve piperidine treatment during Fmoc-SPPS than Dde. ivDde has been applied in de Visser's synthesis of the Dab-rich cyclopeptide antibiotic polymyxin B1 and analogs.
Catalog No. 50-194-7153