Learn More
Med Vet International The Lone Star Veterinary Retractor System, Ring Retractor Only, Large, J4405
Supplier:  Med Vet International JOR4405
Encompass_Preferred
The Lone Star Veterinary Retractor System consists of two components, the autoclavable plastic ring and the accessory elastic stays. Because of its unique design, the retractor ring easily can be adapted to the patient’s anatomy. The elastic stays allow the surgeon flexibility over the amount of tension applied and precise retraction of the incision without obstruction. Prep and drape patient in usual manner, then apply the retractor.
Catalog No. 50-005-84450