Learn More
Perkin Elmer US LLC The OmniCure S2000 UV Lamp
Supplier:  Perkin Elmer US LLC N5365049
Encompass_Preferred
The OmniCure S2000 UV Lamp is a replacement lamp for the OmniCure S2000 Photo Accessory (N5330612). It is a high pressure 200 W mercury vapor short arc lamp with a guaranteed life of 2000 hours.
Catalog No. 50-211-1448
Only null left
Add to Cart
Request a Quote