Learn More
Bachem Fmoc-Asp(OMpe)-OH
SDP
Supplier:  Bachem 4033988.0001
Encompass_Preferred
Fmoc-Asp(OMpe)-OH, 180675-08-5, C25H29NO6, Mr 439.51, 1g. The OMpe (O-3-methylpent-3-yl) side chain protecting group significantly reduces aspartimide formation, an undesired side-chain reaction encountered with the repetitive piperidine treatment needed for Fmoc removal.
Catalog No. 50-194-7124